Medicom™ SafeBasics™ Self-Sealing Sterilization Pouches
SafeBasics™ Self-Sealing Sterilization Pouches with dual indicators to confirm sterilization.
Supplier: Medicom™ 99010
Description
- Self-sealing sterilization pouches made from high-quality materials to deliver reliable sterilization processing at an affordable price.
- Dual (internal/external) steam indicators provide reliable confirmation of sterilization.
Specifications
Autoclavable | |
Green | |
Paper | |
3.5 x 9 in. |
Self-adhesive | |
Single-use | |
Non-sterile |