Greiner Bio-One Safety Blood Collection/Infusion Set without Luer
Specially developed for blood collection from patients with difficult vein conditions
Supplier:  Greiner Bio-One 450091
Catalog No. 22-041-122
Explore available promotions
Description
Description
  • Simple activation of the safety mechanism via lightly pressing in both sides of the hub
  • Transparent view window provides clear vein entry indication
  • Complete activation of the safety mechanism with an audible click
Specifications
Specifications
Blood Collection Set
21 G
3/4 in.
19 cm