missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Safety Blood Collection Set
Sterile and preassembled for ease of use
Supplier: Greiner Bio-One 450184
Description
- Cost effective; ensures your patient only has to undergo one venipuncture for all procedures
- Allows collection of blood into blood culture bottles and Vacuette™ blood collection tubes in one procedure
- Safety Blood Collection Set with preattached holder has been designed for use with most commonly used blood culture bottles
- Compatible with BacT/Alert and BacTec blood culture bottles
- Standard Blood Collection Set (without safety machanism) is also available with preassembled blood culture holder; contact your Fisher HealthCare Representative
Specifications
11.81 in. | |
Sterile | |
Yes | |
Clear | |
CE, ISO 11014 |
30 cm | |
Safety Blood Collection Set | |
21 G | |
Blood culture bottles |