missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Safety-Labeled Wash Bottles
Includes Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols. Fisherbrand™ Safety-Labeled Wash Bottles also include Chemical Abstract Service (CAS) number, protective equipment symbols and target organ inf.
$111.58 - $147.36
Specifications
Capacity (Metric) | 500 mL |
---|---|
Capacity (English) | 17 oz. |
Color | Natural |
Material | Polyethylene |
Certifications/Compliance | OSHA 29 CFR 1910.1200 |
Catalog Number | Mfr. No. | Chemical Label | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Chemical Label | Price | Quantity | ||||||
FB0340923M
|
Fisherbrand™
FB0340923M |
Deionized Water |
Pack of 6 Each for $118.82
Case of 4 Pack for $475.28
|
|
||||||
FB0340923N
|
Fisherbrand™
FB0340923N |
Unlabeled |
Pack of 6 Each for $147.36
Case of 4 Pack for $589.44
|
|
||||||
FB0340923G
|
Fisherbrand™
FB0340923G |
Water |
Pack of 6 Each for $111.58
Case of 4 Pack for $446.32
|
|
||||||
FB0340923K
|
Fisherbrand™
FB0340923K |
Distilled Water |
Pack of 6 Each for $118.82
Case of 4 Pack for $475.28
|
|
||||||
Description
- Easy-to-read printed bottles
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Polypropylene closures
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Use only with solvents compatible with LDPE.
Specifications
500 mL | |
Natural | |
OSHA 29 CFR 1910.1200 | |
Wide | |
53 mm |
17 oz. | |
Polyethylene | |
Not Autoclavable | |
Polypropylene |