Accuform™ Safety Sign, DANGER USE LOCKOUT BEFORE WORKING ON EQUIPMENT
Signs identify and indicate the potential, type, and degree of hazards that may lead to accidental injury or property damage. They comply in accordance with OSHA 29 CFR 1910.145 and ANSI Z535 requirements
Supplier: Accuform™ MLKT021VS
Description
- Printed matte surface has permanent acrylic adhesive back
- Flexible vinyl can be applied to curved or uneven surfaces
- Moisture resistant for a various light-duty indoor and sheltered outdoor marking needs
- Square corners and scored back liner for quick peel-off and application
- 7 x 10, Adhesive Vinyl
Specifications
Vinyl | |
Self-adhesive | |
English | |
Red on White |
7 x 10 in. | |
Rectangular | |
DANGER USE LOCKOUT BEFORE WORKING ON EQUIPMENT |