missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Accuform™ Safety Sign - STOP FACE MASK REQUIRED EVEN IF YOU HAVE BEEN VACCINATED, Aluminum
Accuform™ vaccination signs communicate health and safety policies, reinforce procedures, and identify locations
$29.27 - $48.59
Specifications
Mounting Type | Corner Holes |
---|---|
Shape | Rectangular |
Languages | English |
Displayed Text | STOP FACE MASK REQUIRED EVEN IF YOU HAVE BEEN VACCINATED |
Color | Blue |
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
19-119-113
|
Accuform™
MBDX909VA |
Each for $29.27
|
|
||||||
19-119-116
|
Accuform™
MBDX910VA |
Each for $48.59
|
|
||||||
Description
Specifications
Corner Holes | |
English | |
Blue |
Rectangular | |
STOP FACE MASK REQUIRED EVEN IF YOU HAVE BEEN VACCINATED |