missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ SafTest™ Test Kit

SafTest™ Test Kit measures percentage analytes in given samples. Choose from 6 different kits for use with SafTest instrument and consumable platforms and detection kits.

$1164.52 - $1590.15

Specifications

Detection Method Bichromatic Photometer
No. of Reactions 100 Tests
Quantity 1 Ea.
Storage Requirements 2°C to 8°C
View More Specs

Includes: Reagent A, Reagent B, Reagent C, Calibrators

Products 6
Catalog Number Mfr. No. Target Molecule Price Quantity  
Catalog Number Mfr. No. Target Molecule Price Quantity  
MP7KTAK2000
View Documents
MP Biomedicals™
07KTAK2000
Alkenal
Each for $1,552.69
 
MP7KTFA2000
View Documents
MP Biomedicals™
07KTFA2000
Free Fatty Acid
Each for $1,164.52
 
MP7KTMA2000
View Documents
MP Biomedicals™
07KTMA2000
Malonaldehyde
Each for $1,547.31
 
MP7KTPF2000
View Documents
MP Biomedicals™
07KTPF2000
Percent Fat
Each for $1,590.15
 
MP7KTPR2000
View Documents
MP Biomedicals™
07KTPR2000
Peroxide
Each for $1,429.13
 
MP7KTTG2000
View Documents
MP Biomedicals™
07KTTG2000
Total Glycerin
Each for $1,590.15
 
Description

Description

MP Biomedicals' Test Kits contain all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.

  • Choose from 6 different kits
  • For use with SafTest instrument and consumable platforms
  • Results in as little as 30 minutes
  • Detection method: Bichromatic Photometer
  • Test Kits contain all reagents and calibrators necessary to run samples.
  • A control is also included in each Test Kit.
  • Each kit will allow for testing of approximately 100 samples.
  • Sample dilution will likely be required for appropriate analysis.
    • Alkenal Test Kit
      • Research Area: Lipid degradation and oxidation
      • Contains all reagents, calibrators
    • Free Fatty Acid Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analyte(s)
      • Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.1 - 2.01% free fatty acid
    • Malonaldehyde Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.32 mg/kg of sample
    • Percent Fat Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Calibrators, Control Analytes(s)
      • Material Required: SafTest Instrument Platform
      • Quantitation Range: 0.06 to 0
    • Peroxide Test Kit
      • Research Area: Lipid degradation and oxidation
      • Components: Reagent A, Reagent B, Reagent C, Calibrators, Control Analyte(s)
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.00 to 0.200 MEQ/kg of sample
    • Total Glycerin Test Kit
      • Research Area: Biodiesel
      • Components: Reagent A, Reagent B, Calibrators, Control
      • Materials Required: SafTest Instrument Platform, Consumables Platform and Dispenser Set
      • Quantitation Range: 0.008 to 0.06% of sample
Specifications

Specifications

Bichromatic Photometer
100 Tests
1 Ea.
2°C to 8°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.