missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saint Gobain Life Sciences Chemware™ Bendable PTFE Stirring Rod DFS Item

Chemware™ Bendable PTFE Stirring Rod contains a fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers.

Supplier:  Saint Gobain Life Sciences D1069091

 View more versions of this product

Catalog No. 14-518-10F


Explore available promotions

Description

Description

  • Heat resistant to 288°C (550°F)
  • Completely inert to highly corrosive reagents
  • Anti-stick properties facilitate cleaning
  • Unbreakable
Specifications

Specifications

Fully enclosed steel insert
16 in.
Stirring Rod
PTFE
8 mm
0.3 in.
1 Ea.
40 mm
Videos
SDS
Documents

Documents

Promotions

Promotions