Learn More
Thermo Scientific™ SalI (10 U/μL)
The SalI restriction enzyme recognizes G^TCGAC sites and cuts best at 37°C in O buffer.
Supplier: Thermo Scientific™ ER0642
Description
Lambda DNA digested with SalI, 0.7% agarose, 2 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'...G▵T C G A C...3'3'...C A G C T▵G...5'
Conditions for 100% Activity
- 1X Buffer O:50mM Tris-HCl (pH7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer
- SalI is supplied in:10mM Tris-HCl (pH7.4 at 25°C),100mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Ligation and Recleavage
- After 50-fold overdigestion with SalI, more than 95% of the DNA fragments can be ligated and recut
Methylation Effects
- Dam: never overlaps — no effect
- Dcm: never overlaps — no effect
- CpG: completely overlaps — blocked
- EcoKI: never overlaps — no effect
- EcoBI: never overlaps — no effect
Digestion of Agarose-embedded DNA
- Minimum 5units of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16hours
Compatible Ends
- Eco88I, SmoI, XhoI
Note:
Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity. Supercoiled forms of pBR322 and pUC require 10-fold overdigestion with SalI to achieve complete digestion. Incubation at 25°C results in 50-75% activity.
Specifications
10 U/μL | |
Sal I | |
Yes | |
No | |
Restriction Enzyme |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X Buffer O | |
37°C | |
5 x 1,500 units | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.