missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salicin, MP Biomedicals

Salicin is a substrate for β-glucosidase

Supplier:  MP Biomedicals Inc 0210283310

 View more versions of this product

Catalog No. ICN10283310


Explore available promotions

Description

Description

Specifications

Specifications

Salicin
C13H18O7
10 g
NGFMICBWJRZIBI-UJPOAAIJSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
439503
286.3
138-52-3
MFCD00006590
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
286.28
CHEBI:17814
Powder

Chemical Identifiers

138-52-3
286.28
NGFMICBWJRZIBI-UJPOAAIJSA-N
439503
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C13H18O7
MFCD00006590
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
CHEBI:17814
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 205-331-6

SDS
Documents

Documents

Promotions

Promotions