Learn More
Salicylaldehyde azine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1052718
Description
Salicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSalicylaldehyde azine is used in the preparation of 2-oxo-2H-1-benzopyran-3-carboxylic acid 2-(2-oxo-2H-1-benzopyran-3-carbonyl)hydrazide by reacting with propadienedione . It has anthelmintic activity.
Solubility
Soluble in dimethyl sulfoxide, methanol and ethyl acetate. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Salicylaldehyde azine | |
216°C to 222°C | |
C14H12N2O2 | |
50 g | |
salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone | |
SPEXYYIULCBQJR-UHFFFAOYSA-N | |
6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one | |
6849893 | |
97% |
959-36-4 | |
97% | |
MFCD00043496 | |
1913597 | |
Soluble in dimethyl sulfoxide,methanol and ethyl acetate. Slightly soluble in water. | |
C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1 | |
240.262 | |
240.26 |
Chemical Identifiers
959-36-4 | |
240.262 | |
SPEXYYIULCBQJR-UHFFFAOYSA-N | |
6849893 | |
C1=CC(=CNNC=C2C=CC=CC2=O)C(=O)C=C1 |
C14H12N2O2 | |
MFCD00043496 | |
salicylaldehyde azine, salicylalazine, salicylaldazine, benzaldehyde, 2-hydroxy-, 2-hydroxyphenyl methylene hydrazone, 2-hydroxybenzaldehyde azine, 2-1e-e-2-2-hydroxyphenyl methylidene hydrazin-1-ylidene methyl phenol, 2,2'-1,2-hydrazinediylidenedi methylylidene diphenol, benzaldehyde, 2-hydroxy-, 2-2-hydroxyphenyl methylene hydrazone, 1,2-bis e-salicylidene hydrazine, 6-6-oxo-1-cyclohexa-2,4-dienylidene methylhydrazo methylidene-1-cyclohexa-2,4-dienone | |
6-[[2-[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]hydrazinyl]methylidene]cyclohexa-2,4-dien-1-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 213-500-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only