missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, 99%

CAS: 65-45-2 | C7H7NO2 | 137.14 g/mol

$253.57 - $253.57

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007978
InChI Key SKZKKFZAGNVIMN-UHFFFAOYSA-N
Synonym salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
PubChem CID 5147
ChEBI CHEBI:32114
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)C1=CC=CC=C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229360010
SDS View Documents
Thermo Scientific Chemicals
229360010
1 kg Plastic bottle
Each for $253.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65-45-2
137.14
SKZKKFZAGNVIMN-UHFFFAOYSA-N
5147
2-hydroxybenzamide
C7H7NO2
MFCD00007978
salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
CHEBI:32114
NC(=O)C1=CC=CC=C1O

Specifications

65-45-2
White
98.5% min. (on anhydrous substance)
C7H7NO2
MFCD00007978
10, 87
salicylamide, o-hydroxybenzamide, benzamide, 2-hydroxy, salicylic acid amide, 2-carbamoylphenol, flarpirina, morsarinas, algamon, algiamida, allevin
SKZKKFZAGNVIMN-UHFFFAOYSA-N
2-hydroxybenzamide
5147
137.14
Crystalline Powder
139.0°C to 142.0°C
Authentic
Plastic bottle
C6H4(OH)CONH2
1 kg
15, 8465
Solubility in water: soluble in hot water. Other solubilities: 0.2% in water (30°C), 10.0% in propylene glycol (5°C), soluble in alcohol, chloroform and ether, slightly soluble in carbon tetrachloride
NC(=O)C1=CC=CC=C1O
137.14
CHEBI:32114
99%
Salicylamide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-609-3

RTECSNumber : VN6475000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only