missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
![](http://beta-static.fishersci.ca/images/Gray.gif)
Salicylic Acid (Crystalline Powder/Certified ACS), Fisher Chemical
Supplier: Fisher Chemical FLA277500
Specifications
Salicylic Acid | |
158°C | |
211°C | |
C7H6O3 | |
MFCD00002439 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
2-hydroxybenzoic acid | |
338 | |
138.12 | |
Certified ACS | |
0.010% max. | |
Solid |
2.4 | |
69-72-7 | |
White | |
2-HOC6H4COOH | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
OC(=O)C1=CC=CC=C1O | |
138.12 | |
CHEBI:16914 | |
≥99.0% | |
500 g | |
Poly Bottle | |
0.3mbar at 95°C |
Chemical Identifiers
69-72-7 | |
138.12 | |
YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
338 | |
2-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002439 | |
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
CHEBI:16914 | |
OC(=O)C1=CC=CC=C1O |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Gray.gif)