Learn More
Salicylsalicylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 133400250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Salicylsalicylic acid | |
552-94-3 | |
97.5% min. (HPLC) | |
C14H10O5 | |
25g | |
15,8477 | |
Solubility in water: hydrolysis. Other solubilities: soluble in alcohol and ether, sparingly soluble in benzene, slightly soluble in acetone | |
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O | |
258.22 | |
CHEBI:9014 | |
98% |
99% | |
Authentic | |
Glass bottle | |
MFCD00020252 | |
10,II,54 | |
salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
WVYADZUPLLSGPU-UHFFFAOYSA-N | |
2-(2-hydroxybenzoyl)oxybenzoic acid | |
5161 | |
258.22 |
Chemical Identifiers
552-94-3 | |
258.22 | |
WVYADZUPLLSGPU-UHFFFAOYSA-N | |
5161 | |
2-(2-hydroxybenzoyl)oxybenzoic acid |
C14H10O5 | |
MFCD00020252 | |
salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
CHEBI:9014 | |
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 209-027-4
RTECSNumber : DH2080000
TSCA : TSCA
RUO â Research Use Only