missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Salvage Poly Drum
Salvage drums are designed for use with damaged or leaking containers of hazardous materials, or for used absorbent materials contaminated with hazardous substances.
Supplier: Eagle™ 1654
Description
- For use with damaged or leaking containers of hazardous materials or used absorbent materials contaminated with hazardous substances
- Salvage Drum labeling
- Passed a 3-PSI pressure test
Specifications
Salvage Drum | |
Metal Lever-lock | |
75.8 L | |
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
19 in. | |
16 lb. |
Yes | |
20 gal. | |
Yellow | |
HDPE | |
48.3 cm |