missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium, 99.9%, (trace metal basis), 20 to 40 mesh, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 294780100

 View more versions of this product

Catalog No. AC294780100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

1778.0°C
1072.0°C
7440-19-9
Powder
99.9%
Sm
3132
15, 8484
Solubility in water: reacts. Other solubilities: forming hydrogen
[Sm]
150.36
CHEBI:33374
99.9%
Plastic bottle
Samarium
40 Mesh
Gray
10 g
Sm
MFCD00011233 MFCD00151299
17,305
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samarium
23951
150.36
Trace Metal Basis

Chemical Identifiers

7440-19-9
150.36
KZUNJOHGWZRPMI-UHFFFAOYSA-N
23951
samarium
Sm
MFCD00011233 MFCD00151299
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
CHEBI:33374
[Sm]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gas.
Flammable solid.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a dry place.
Store in a closed container.
In case of fire: Use dry sand,dry chemical or alcohol-resistant foam for extinction.

GHS Signal Word: Danger

EINECSNumber : 231-128-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only