Learn More
Samarium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035448FF
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Spécifications
Silver | |
0.025mm (0.001 in.) thick | |
(REO) | |
MFCD00011233 MFCD00151299 | |
25 x 25 mm | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
samarium | |
23951 | |
99.9% | |
Mylar under Argon | |
Foil |
Samarium foil | |
7440-19-9 | |
Sm | |
UN3178 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
[Sm] | |
150.36 | |
CHEBI:33374 | |
Odorless | |
≈0.12g/25x25mm |
Identifiants chimiques
7440-19-9 | |
150.36 | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
23951 | |
samarium |
Sm | |
MFCD00011233 MFCD00151299 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
CHEBI:33374 | |
[Sm] |
Sécurité et manipulation
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260
missing translation for 'dotInformation' : Hazard Class: 4.3; Packaging Group: III
missing translation for 'einecsNumber' : 231-128-7
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only