missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 043217FF

 View more versions of this product

Catalog No. AA43217FF


Explore available promotions

Description

Description

Used in control rods in nuclear reactors, as a catalyst in organic chemistry, a pyrophoric metal in lighter flints, and a core in carbon arc-lamp electrodes. Also used in alloys with cobalt to make strong permanent magnets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
Specifications

Specifications

Samarium foil
7440-19-9
Sm
25 x 25 mm
Insoluble in water.
[Sm]
150.36
CHEBI:33374
Odorless
≈4.9g/25x25mm
1.0mm (0.04 in.) thick
(REO)
MFCD00011233 MFCD00151299
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samarium
23951
99.9%
Mylar under Argon
Foil

Chemical Identifiers

7440-19-9
150.36
KZUNJOHGWZRPMI-UHFFFAOYSA-N
23951
samarium
Sm
MFCD00011233 MFCD00151299
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
CHEBI:33374
[Sm]
Videos
Safety and Handling

Safety and Handling

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Hazard Class: 4.3; Packaging Group: III

EINECSNumber : 231-128-7

TSCA : Yes

storageNote1 :

  • Packaged in mylar under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only