missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Samarium(II) iodide, 0.1M solution in THF, stabilized with Samarium powder, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 294991000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Samarium(II) iodide, 0.1M solution in THF | |
Blue-Green | |
Solution | |
I2Sm | |
MFCD00058873 | |
15, 8486 | |
404.17 | |
AcroSeal™ Glass Bottle | |
0.9220g/mL |
0.1M solution in THF, Stabilized with samarium powder | |
32248-43-4,109-99-9 | |
100 mL | |
SmI2 | |
−17°C | |
404.17 | |
0.1M, exact strength on the certificate of analysis | |
0.922 |
RUO – Research Use Only