Learn More
Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
$149.81 - $484.04
Chemical Identifiers
CAS | 13759-83-6 |
---|---|
Molecular Formula | H12N3O15Sm |
Molecular Weight (g/mol) | 444.46 |
MDL Number | MFCD00149857 |
InChI Key | HDCOFJGRHQAIPE-UHFFFAOYSA-N |
Synonym | samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate |
PubChem CID | 203081 |
SMILES | O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC316150250
|
Thermo Scientific Chemicals
316150250 |
25 g | Plastic bottle |
Each for $149.81
|
|
||||
AC316151000
|
Thermo Scientific Chemicals
316151000 |
100 g | Plastic bottle |
Each for $484.04
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
13759-83-6 | |
444.46 | |
HDCOFJGRHQAIPE-UHFFFAOYSA-N | |
203081 |
H12N3O15Sm | |
MFCD00149857 | |
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate | |
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
Yellow | |
Crystalline Powder and/or Crystals | |
99.9% | |
Sm(NO3)3·6H2O | |
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate | |
HDCOFJGRHQAIPE-UHFFFAOYSA-N | |
samarium(3+);trinitrate;hexahydrate | |
203081 | |
99.9% | |
Plastic bottle |
13759-83-6 | |
25 g | |
H12N3O15Sm | |
MFCD00149857 | |
Solubility in water: soluble | |
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
444.46 | |
444.5 | |
Trace Metal Basis | |
Samarium(III) nitrate hexahydrate |
Safety and Handling
GHS H Statement:
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only