missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals

$193.84 - $815.96

Chemical Identifiers

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40297HF
View Documents
Thermo Scientific Chemicals
040297HF
10 mm
Each for $193.84
 
AA40297DH
View Documents
Thermo Scientific Chemicals
040297DH
25 mm
Each for $302.18
 
AA40297DJ
View Documents
Thermo Scientific Chemicals
40297DJ
50 mm
Each for $538.53
 
AA40297DM
View Documents
Thermo Scientific Chemicals
040297DM
100 mm
Each for $815.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

7440-19-9
150.36
KZUNJOHGWZRPMI-UHFFFAOYSA-N
23951
samarium
Sm
MFCD00011233 MFCD00151299
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
CHEBI:33374
[Sm]

Specifications

7440-19-9
10 mm
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samarium
23951
99.9%
Mylar under Argon
(metals basis excluding Ta)
Cast Rod
Sm
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
[Sm]
150.36
CHEBI:33374
Odorless
Samarium rod, 6.35mm (0.25 in.) dia.
Videos
Safety and Handling

Safety and Handling

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Hazard Class: 4.3; Packaging Group: III

EINECSNumber : 231-128-7

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only