missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Samco™ Graduated Transfer Pipettes

Measure more than one volume accurately by using these graduated polyethylene transfer pipettes. Built-in pipette bulb eliminates need for plugging, packaging, and autoclaving.

Supplier:  Thermo Scientific™ 28220S

 View more versions of this product

Catalog No. 02-708-014


Explore available promotions

Description

Description

Accurately measure more than one volume by using Thermo Scientific™ Samco™ Graduated Transfer Pipettes. Single-unit design features a built-in pipette bulb that eliminates need for plugging, packaging, and autoclaving while preventing cross-contamination.

  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
Specifications

Specifications

0.8 mL
3.0 mm
Sterile
Yes
20/Bag, 500/Pk., 4000/Cs.
11.6 cm
Single-use
Transfer Pipette, Pediatric, Graduated to 0.1mL, 0.2mL, 0.3mL
LDPE
22
0.1, 0.2, 0.3 mL
1.5 mL
Graduated Transfer Pipet
Videos
SDS
Documents

Documents

Promotions

Promotions