missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers

Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.

$700.16 - $700.16

Specifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Shape Cylindrical
Sterility Non-sterile
View More Specs

Products 1
Catalog Number Mfr. No. Closure Color Price Quantity  
Catalog Number Mfr. No. Closure Color Price Quantity  
13-711-86
View Documents
Thermo Scientific™
040506
Yellow
Case of 600 Each for $700.16
 
Description

Description

Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:

  • Available nonsterile or gamma irradiation sterilized
  • Available labled or non-labled
  • Available bulk or individually packed
  • In cases of 500
Specifications

Specifications

Specimen Container
Virgin Polypropylene
Non-sterile
48 mm
Cap and container bagged separately
600/Cs.
Yes
Cylindrical
40 mL
Bio-Tite Lid
48 mm
600/Pk.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'