MilliporeSigma™ Supelco™ Sample Loop for Model 7725 Injector, Stainless Steel
Sample loop for model 7725 injector (volume 100 μL) is best suited for advanced fluid-handling solutions for broad range of analytical instrumentation and in vitro diagnostic systems. It is useful in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Supplier: MilliporeSigma™ Supelco™ SGMA57624
Specifications
Stainless Steel | |
Model 7725 | |
10 μL |