Cytiva Sample Valve
Supplier:  Cytiva 29023896
Catalog No. 36-104-1205
Explore available promotions
Description
Description
For use with the AKTA start automated protein purification system. The sample valve allows either the buffer or the sample to enter the flow path. The sample valve enables the direct application of the sample onto the column using the pump.
Specifications
Specifications
Enables the direct application of the sample onto the column using the pump
AKTA start Automated Protein Purification System