Learn More
ITL BioMedical SampLok™ Sampling Kit DFS Item
SampLok™ Sampling Kit provides a more straightforward, safer, and efficient way to collect and transfer platelet samples for bacterial testing.
Supplier:  ITL BioMedical A100754
 Afficher plus de versions de ce produit
Numéro de catalogue. 22-799-909
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Reduces the number of procedure steps, streamlining and simplifying the collection and transfer of biological samples
  • SampLok™ Sampling Kit can be used with various culture bottles and vacuum tubes
  • Protect staff from needlestick injuries
Spécifications
Spécifications
Collection and Transport System
Sterile
Culture Bottles and Vacuum Tubes
20 mL
Tube
Inegrated Safety Lid
110 per Case
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS