missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sand, Ottawa (For Cement Testing), Fisher Chemical™

ASTM C778

Supplier:  Fisher Chemical S2350

 View more versions of this product

Catalog No. S23-50


Explore available promotions

Description

Description

Specifications

Specifications

2230°C
1610°C
Beige
Powder/Solid
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.08
CHEBI:30563
Fiber Drum
Sand, Ottawa
For cement testing
14808-60-7
50 kg
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
Pass Test
2.65g/cm³

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
silanedione
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Cancer hazard. May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Do not breathe dust. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:0
Instability:0

SDS Documents

Documents

Accessories
Promotions

Promotions