missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sand, White Quartz, Honeywell Fluka™

50-70 mesh particle size

$161.15 - $498.25

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002246
View Documents
Honeywell Chemicals
274739500G
500 g
Each for $161.15
 
6002245
View Documents
Honeywell Chemicals
2747391KG
1 kg
Each for $200.80
 
6002247
View Documents
Honeywell Chemicals
2747395KG
5 kg
Each for $498.25
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563

Specifications

White
500 g
SiO2
NONH for all modes of transport
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08g/mol
14808-60-7
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.08
CHEBI:30563
Sand, White Quartz
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-878-4

RTECSNumber : VV7330000

SDS
missing translation for 'documents'

missing translation for 'documents'