Learn More
Applied Biosystems™ Sanger Sequencing Kit
Sanger Sequencing Kit includes all reagents needed for sample preparation for sanger sequencing
Supplier: Applied Biosystems™ A38073
Includes: 2 x 100 rxns ExoSAP-IT Express, 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), 1 x 800 μL BigDye Terminator v3.1 ready reaction mix, 1 tube M13 (-21) primer, 1 tube pGEM control DNA, 1 x 2 mL XTerminator solution, 1 x 9 mL SAM solution
Specifications
| • 2 x 100 rxns ExoSAP-IT Express, store at -5 to -30°C • 2 x BigDye Terminator v1.1 and v3.1 sequencing buffer (5X), store at 2–8°C • 1 x 800 µL BigDye Terminator v3.1 ready reaction mix, store at -15 to -25°C • 1 tube M13 (-21) primer, store at -15 to -25°C • 1 tube pGEM control DNA, store at -15 to -25°C • 1 x 2 mL XTerminator solution, store at 2–8°C • 1 x 9 mL SAM solution, store at room temperature or 4°C |
|
| Sequencing Instruments (Capillary Electrophoresis-Based) | |
| 200 | |
| 200 Reactions |
| Kit | |
| Standard Sequencing | |
| Sanger Sequencing Kit | |
| Approved for shipment at Room Temperature or on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.