missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Santonin 98.0+%, TCI America™

Supplier:  TCI America S052125G

 View more versions of this product

Catalog No. S052125G


Explore available promotions

Description

Description

Specifications

Specifications

Santonin
173°C
C15H18O3
25 g
XJHDMGJURBVLLE-BOCCBSBMSA-N
(3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
221071
246.31
Crystalline Powder
481-06-1
White
MFCD00135865
santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum
C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O
246.31
CHEBI:16363
≥98.0% (HPLC,T)

Chemical Identifiers

481-06-1
246.31
XJHDMGJURBVLLE-BOCCBSBMSA-N
221071
(3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
C15H18O3
MFCD00135865
santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum
CHEBI:16363
C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LE3150000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions