missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Santonin | |
173°C | |
C15H18O3 | |
5 g | |
XJHDMGJURBVLLE-BOCCBSBMSA-N | |
(3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione | |
221071 | |
246.31 | |
Crystalline Powder |
481-06-1 | |
White | |
MFCD00135865 | |
santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum | |
C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O | |
246.31 | |
CHEBI:16363 | |
≥98.0% (HPLC,T) |
Chemical Identifiers
481-06-1 | |
246.31 | |
XJHDMGJURBVLLE-BOCCBSBMSA-N | |
221071 | |
(3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione |
C15H18O3 | |
MFCD00135865 | |
santonin, alpha-santonin, --alpha-santonin, santoninic anhydride, --santonin, unii-1vl8j38ero, semenen, --santonine, 1vl8j38ero, santoninum | |
CHEBI:16363 | |
C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O |
Safety and Handling
RTECSNumber : LE3150000
TSCA : Yes