missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Saponin, pract., from Quillaja Saponaria Molina

CAS: 74499-23-3

$696.50 - $3069.36

Chemical Identifiers

CAS 74499-23-3
MDL Number MFCD00081981
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC419235000
SDS View Documents
Thermo Scientific Chemicals
419235000
500 g Glass bottle
Each for $696.50
Only null left
 
AC419230025
SDS View Documents
Thermo Scientific Chemicals
419230025
2.5 kg Plastic drum
Each for $3,069.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

74499-23-3
MFCD00081981

Specifications

74499-23-3
Red to Tan
Authentic
MFCD00081981
14, 8365
Powder
(c=20, water) (1 cm cell), (c=20, water) (1 cm cell) at 0nm, 2 max. at 520nm
3.8 to 4.6 (20% soln.)
Glass bottle
500 g
Solubility in water: soluble. Other solubilities: freely soluble in diluted alcohol
Saponin, pract.
Videos
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only