missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Sarcosine, 98%

Supplier:  Thermo Scientific Chemicals 157741000

 View more versions of this product

Catalog No. AC157741000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sarcosine
107-97-1
100 g
Plastic bottle
C3H7NO2
CH3NHCH2CO2H
N-Methylglycine
CNCC(O)=O
89.09
CHEBI:15611
≥97.5% (total base)
Colorless or White to Yellow
Crystals or Powder
Authentic
98%
2% max.
MFCD00004279
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
2-(methylamino)acetic acid
1088
89.09

Chemical Identifiers

107-97-1
89.09
FSYKKLYZXJSNPZ-UHFFFAOYSA-N
1088
2-(methylamino)acetic acid
C3H7NO2
MFCD00004279
N-Methylglycine
CHEBI:15611
CNCC(O)=O
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only