Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
Supplier:  Sartorius 16417
 View more versions of this product
Catalog No. 14-555-780
Explore available promotions
Description
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780
  • Biosart 100 Membrane Lifter
  • For easy transfer of the membrane onto agar
Adapter, Polypropylene - 14-555-779
  • Biosart 100 Adapter
  • Fits Biosart 100 Monitor onto 50 mm supports
Adapter, Silicone - 14-555-778
  • Biosart 100 Adapter
  • 50 mm stainless-steel frit and silicone gasket
  • Fits Biosart 100 Monitor onto 16840 or 16841 base
Adapter, Stainless Steel - 14-555-781
  • Biosart 100 Adapter to accommodate various funnel types
  • Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
Stopper, Silicone - 14-555-817
  • Replacement stopper for 16385
  • 16385 adapts Biosart 100 Monitors on Combisart manifold
Specifications
Specifications
Reusable
ABS Copolymer
For easy transfer of the membrane onto agar
Sartorius Biosart 100
Non-sterile
Adapter