Sartorius Accessories for the MD8 Airscan™ Air Sampler
Accessories for use with the MD8 Airscan™ Air Sampler. The MD8 is used for quantitative detection of airborne organisms, mainly in sterile areas of Class A/B cleanrooms, isolators, or blow-fill-seal machines.
Supplier:  Sartorius 17801
 View more versions of this product
Catalog No. 14-555-884
Explore available promotions
Description
Description
Accessories
Adapter, Connector - 14-555-869
  • Connects airscan to isolator via flexible PVC hose and filter capsule
  • DN 30 hose barb to 1 to 11 in. sanitary flange
Adapter, Connector - 14-555-890
  • Connects airscan to isolator via silicone tubing and filter capsule
  • DN 25 hose barb to 1 to 11 in. sanitary flange
Adapter, Plate - 14-555-876
  • For culture media plates
Adapter, Ring - 14-555-912
  • Replacement part for aluminum stack
  • Intermediate (middle) part
Aluminum Stack - 14-555-878
  • Stack of 10 open filter holders
Calibration Kit - 14-555-868
  • Portable flowrate measuring device for the MD8 series air samplers
Capsule, Sartoflor - 14-555-889
  • With PTFE membrane and sanitary flange inlet and outlet
  • For sterile air filtration inserted between the MD8 airscan and isolator
Carrying Case - 14-555-873
  • Stable case for the transport and the storage of unit and accessories
Clamp, Flange - 14-555-891
  • For 1 to 11 in. sanitary flanges
Connector - 14-555-879
  • Aluminum
  • Set of connectors for MD 8 (SM 16743/44)
Connector - 14-555-909
  • Aluminum
  • Connects air sampler inlet to flexible hose
Connector - 14-555-910
  • Aluminum
  • Connects flexible hose to filter holder/adapter
Connector - 14-555-911
  • Flexible hose to filter holder/adapter (S-F)
  • For triclamp
Filter Holder - 14-555-880
  • Holds 80 mm gelatin filters
  • Replacement part for the aluminum stack
Filter Holder - 14-555-884
  • Adapter for gelatin filter units (gelatin disposables) to fit on the MD8 inlet
O-Ring - 14-555-922
  • 7.5 x 1.5 mm
Remote Control - 14-555-877
  • For use with PC (dialog system software)
Specifications
Specifications
Reusable
Aluminum
Filter Holder
MD8 Airscan Air Sampler
Adapter for Gelatin Disposables to Fit on the MD8 Inlet
Product Suggestions
Product Suggestions