missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sartorius Vivaspin™ Filtrate Centrifugal Concentrators

Remove proteins from difficult to filter samples, even whole blood. Sartorius Vivaspin™ Filtrate performs centrifugal ultrafiltration against the direction of centrifugal force for fast and simple preparation of protein-free ultrafiltrates. PES and CTA membranes for 5000 to 300,000 Dalton MWCO.

Supplier:  Sartorius 13269E

 View more versions of this product

Catalog No. 14-558-223


Explore available promotions

Description

Description

Offered with multiple MWCOs and two membrane options:

  • Cellulose triacetate (CTA) features high hydrophilicity and very low non-specific binding.
    • Cast without a support that could bind filtered solutes, these membranes are ideal where high target molecule recovery from the filtrate is important.
  • Polyethersulfone (PES) membranes exhibit low fouling characteristics, exceptional flux and broad pH compatibility.
    • Have a uniform surface, devoid of hydrophobic or hydrophillic interactions, offering excellent protein recoveries for most solutions.
  • Typical applications for Vivaspin™ Filtrate include:
    • Sample preparation for free drug
    • Metabolite or hormone analyses
    • Sample deproteinization
    • Separation of proteins from low molecular weight substances in biological samples
  • Sample capacity of 0.5 to 2.5 mL
  • May be used in fixed angle or swing bucket rotors, accepting standard conical or flat bottom 15 mL centrifuge tubes
  • Typically filter 90% of the initial sample volume in 30 minutes
  • Avoid premature membrane blockage by particle loaded samples
  • Retrieve the ultrafiltrate immediately after centrifugation without any disassembly
Specifications

Specifications

Reusable
PES
0.5 to 2.5 mL
Small volume centrifugal ultrafiltration of biological samples
12 Pk.
93 mm
Centrifugal Ultrafiltration
100 kDa
Non-sterile
2000xG (Fixed Angle Rotor); 2500xG (Swinging Bucket Rotor)
0.5 to 2.5 mL
14 mm
Videos
SDS
Documents

Documents

Promotions

Promotions