Sartorius Disposable Plastic Syringes
Disposable Plastic Syringes that can be used with all syringe filter holders and 3-way valves.
Supplier: Sartorius 16644E
Description
- Male luer-lok connector
- Comes with needle
- Sterile
Specifications
Plastic | |
For filter holders | |
Single-use | |
Cone | |
Sterile |
19 G | |
Syringe | |
5 mL | |
Male Luer Lock |