Sartorius Granite Platforms for Sartorius Balances
With vibration isolators
Supplier:  Sartorius U121201315
 View more versions of this product
Catalog No. 13-735-736
Explore available promotions
Description
Description
  • Stabilizes weighing results against environmental vibrations
  • Ideal for use with any balance
Specifications
Specifications
CPA Balance, Extended Balance, SE and ME Ultramicro Balance
15 in.
Vibration isolators
Granite
13 in.
Balance Platform