Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
For various Vivacell models
$160.46 - $434.90
Specifications
Type Pressure Head
View More Specs
Products 2
Catalog Number Mfr. No. For Use With Price Quantity  
Catalog Number Mfr. No. For Use With Price Quantity  
14-558-299
View Documents Promotion Details
Sartorius
VCA200
Vivaspin™ 20
Each for $160.46
14-558-303
View Documents Promotion Details
Sartorius
VCA800
Vivacell™ 100
Pack for $434.90
Description
Description
Vivaspin™ 20 is an ultrafiltration unit with unique twin membrane for sample concentration with high process flexibility: centrifugation or centrifugation of pressurized units, as well as pressure filtration in stationary benchtop mode.Vivacell™ 100 is a disposable ultrafiltration unit for the concentration of samples up to 98 mL that can be used either in a centrifuge or for pressure filtration. Vivacell™ 100 thus combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells.
Specifications
Specifications
Pressure Head
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'