Sartorius Stedim Stainless-steel Syringe Filter Holder
Supplier:  Sartorius 16214
 View more versions of this product
Catalog No. 14-555-540
Description
Description
Leak-proof without sealing ring: The top part of the holder can easily be mounted on the bottom part using the enclosed tightening tool. Filter supports in the top and bottom parts allow filtration in both directions.
Specifications
Specifications
Female luer lock (inlet), Male luer slip (outlet)
PTFE-coated Stainless Steel
7 bar
25 mm
Syringe filter holder
3 cm2
<0.1 mL