Sartorius Universal Pump for Sterisart™ Sterility Testing System
Peristaltic pump for sample and media transfer into the Sterisart NF systems - Suitable for isolators
Supplier:  Sartorius 16420
 View more versions of this product
Catalog No. 14-555-827
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The new Sterisart™ Universal Pump is available in two versions: as basic version 16419 and as an upgraded version 16420 with display and user software.The pump can be used in clean rooms, integrated into clean benches, or installed countersunk in the working surface of isolators. Its low, compact design has a space-saving footprint – a great benefit for most clean room benchtops and isolators.
  • closed system – no ventilation for enhanced safety
  • robust and maintenance free
  • compact and ergonomic construction
  • modular design
  • pump available with special software (operator-guided menus; all process sequences can be logged; barcode recognition)
Specifications
Specifications
10.23 in.
Used in clean rooms, integrated into clean benches, or installed countersunk in the working surface of isolators
21 cm
14.6 kg
Display and user software
Stainless Steel, PVC, PET, Silicone
13.22 in.
26 cm
8.26 in.
32.18 lb.
70 to 650 mL/min.
50/60 Hz
100 w
33.6 cm