Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
$285.13 - $2384.65
Catalog Number | Mfr. No. | Description | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | ||||||
36-077-3
|
Thermo Scientific™
SCT120 |
Chemical Trap (order cartridges separately) |
Each for $1,590.88
|
|
||||||
DTK120R
|
Thermo Scientific™
DTK120R |
Chemical Trap Kit for Radioactivity |
Each for $2,174.00
|
|
||||||
36-077-6
|
Thermo Scientific™
DC120R |
Disposable Cartridge (trapping volatile radioactivity) |
Each for $1,224.27
|
|
||||||
20-548-143
|
Thermo Scientific™
DC120A |
Disposable Cartridge (acid neutralization) |
Each for $564.42
|
|
||||||
36-779-3
|
Thermo Scientific™
ANS121 |
Ammonia Neutralizing Solution (Oligo Prep) |
Each for $285.13
|
|
||||||
36-782-0
|
Thermo Scientific™
SCC5 |
CryoCool Heat Transfer Fluid, 5L |
Each for $2,284.29
|
|
||||||
36-079-0
|
Thermo Scientific™
MF190 |
Mist filter replacement cartridge |
Each for $469.27
|
|
||||||
39-792-93
|
Thermo Scientific™
GCF400 |
Glass condensation flask |
Each for $2,384.65
|
|
||||||
Description
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.