Mopec Scalpel Handle for Double Edged Blade
Mopec's double edge blade handle is ideal for trimming surgical specimens and is a versatile tool for any laboratory. The double edge blade is securely held in place, yet easy to replace. The weighted pencil-style handle provides a firm, mobile grip.
Supplier:  Mopec AJ127
Catalog No. AJ127
Explore available promotions
Description
Description
  • Compatible with Mopec AJ111 double edge blades
  • 5 complimentary blades included
Specifications
Specifications
Scalpel Handle
Compatible with Mopec AJ111 Double Edge Blade
Product Suggestions
Product Suggestions