missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium foil, 0.127mm (0.005in) thick, 99.9% (REO), Thermo Scientific Chemicals

Form: Approx. 0.24g/25 x 25mm

$558.81 - $1731.22

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA00294FF
View Documents
Thermo Scientific Chemicals
000294FF
25 x 25 mm
Each for $558.81
 
AA00294FI
View Documents
Thermo Scientific Chemicals
000294FI
50 x 50 mm
Each for $1,731.22
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-20-2
44.96
SIXSYDAISGFNSX-UHFFFAOYSA-N
23952
scandium
Sc
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
CHEBI:33330
[Sc]

Specifications

Silver to Gray
Silver gray foil
Sc
25 x 25 mm
SIXSYDAISGFNSX-UHFFFAOYSA-N
scandium
23952
Odorless
Scandium
7440-20-2
99.9% (REO)
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
[Sc]
44.96
CHEBI:33330
Foil
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-129-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only