missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Scandium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals
$867.44 - $2759.36
Chemical Identifiers
CAS | 7440-20-2 |
---|---|
Molecular Formula | Sc |
Molecular Weight (g/mol) | 44.96 |
MDL Number | MFCD00016323 |
InChI Key | SIXSYDAISGFNSX-UHFFFAOYSA-N |
Synonym | unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted |
PubChem CID | 23952 |
ChEBI | CHEBI:33330 |
IUPAC Name | scandium |
SMILES | [Sc] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-20-2 | |
44.96 | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
23952 | |
scandium |
Sc | |
MFCD00016323 | |
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted | |
CHEBI:33330 | |
[Sc] |
Specifications
7440-20-2 | |
MFCD00016323 | |
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted | |
[Sc] | |
44.96 | |
CHEBI:33330 | |
Odorless | |
≈0.5g/25x25mm | |
Scandium foil, 0.25mm (0.01 in.) thick |
Sc | |
25 x 25 mm | |
SIXSYDAISGFNSX-UHFFFAOYSA-N | |
scandium | |
23952 | |
99.9% | |
Mylar bag | |
Foil | |
(REO) |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-129-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only