ILC Dover SCape CO/CBRN 30 Escape Respirator
Better Protection. Better Fit. Better System. The SCape™ CO/CBRN30 from ILC Dover is the most advanced, FM and NIOSH-approved air-purifying escape respirator on the market.
Supplier: ILC Dover 60001130003
Description
- Provides mission-critical protection from carbon monoxide, chemical, biological, radiological and nuclear contaminants
Specifications
NIOSH | |
Escape Hood | |
Integral 64 lmp blower with indicator light, exhaust valve, one-size neck seal, military grade filter canister, hepa filter, multi layer nylon film lens, battery, clear hood with large 360 vision | |
3/Pk. |