Learn More
MilliporeSigma™ Scepter™ 3.0 Handheld Automated Cell Counter
The Scepter™ 3.0 Handheld Automated Cell Counter provides a fast and convenient method for counting cells or particles using a disposable sensor.
Supplier: MilliporeSigma™ PHCC30000
Description
Using the Coulter Principle, the Scepter™ 3.0 does all the work and rapidly delivers accurate and reliable cell counts.
The Scepter™ 3.0 quantifies cells based on size and discriminates larger cells from smaller debris, unlike vision-based techniques, which rely on object recognition software and cannot reliably detect small cells.
A sample of interest is diluted, then acquired using a precision-engineered, microfluidic Scepter™ 3.0 disposable sensor. In seconds, Scepter™ 3.0 displays cell concentration, cell diameter or volume measurements, along with a histogram of cell distribution.
- Up to 999 histograms can be stored in the instrument.
- The Scepter™ 3.0 has a wireless network interface which allows you to send histograms and data files to your Wi-Fi™ printer or computer easily.
- No more cables.
- The new Charging Station stores Scepter™ 3.0 out-of-theway when not in use, freeing up valuable hood or lab bench space.
Specifications
| Scepter 3.0 Cell Counter | |
| ≥ 100 μL | |
| Acrylonitrile butadiene styrene (ABS) body, polycarbonate digital screen, silicone control buttons | |
| Scepter | |
| 10.9 cm | |
| 28.3 cm | |
| 6.4 cm | |
| Scepter 60 μm or 40 μm Sensors |
| Sensor size: 40 μm (< 40 seconds), 60 μm (< 30 seconds) | |
| Sensor size: 40 μm (5 to 15 μm), 60 μm (8 to 25 μm) | |
| Sensor size: 40 μm (50,000 to 1,500,000 cells/mL), 60 μm (10,000 to 500,000 cells/mL) | |
| 4.3 in. | |
| 11.2 in. | |
| 2.5 in. | |
| Cell counting |
For research use only.