missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Schiff Reagent Solution (Modified), Fisher Chemical
For detection of aldehydes
Supplier: Fisher Chemical SS32500
Specifications
Schiff Reagent Solution | |
632-99-5,7681-57-4,7647-01-0,7732-18-5 | |
1789 | |
Pass Test | |
0.7g/cm³ | |
500 mL |
Modified | |
MFCD00081985 | |
Amber Glass | |
14mmHg | |
Colorless | |
Liquid |