missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scigen O.C.T. Compound Cryostat Embedding Medium

Provides convenient specimen matrix for cryostat sectioning at temperatures of -10°C and below. It is a formulation of 10.24% polyvinyl alcohol, 4.26% polyethylene glycol and 85.5% nonreactive ingredients, being water soluble; it leaves no residue during staining.

Supplier:  Scigen 4586

Catalog No. 23-730-625


Explore available promotions

Description

Description

  • Introduced in 1999, Tissue-Plus O.C.T. Compound has been marketed under several well-known brands worldwide for use as an embedding medium for frozen tissue specimens to ensure optimal cutting temperature
  • Clear, colorless solution, free of particles, water soluble cryostat embedding medium for tissue specimen sectioning and long term cold storage
  • Provides convenient specimen matrix for cryostat sectioning at temperatures of -10°C and below
  • Can be frozen between -10° and -80°C
  • Leaves no residue on slides during staining procedure, eliminating undesirable background staining
  • Provides an exact freeze point resulting in a homogenous matrix along with thinner sections
  • Will not dull cryostat blades
  • Manufactured using ISO 13485 and FDA quality systems
  • Available in convenient 4oz. squeeze bottle

Specifications

Specifications

118 mL
Room temperature
Embedding Media
Cryostat Embedding and Specimen Storage
Clear
10.24% Polyvinyl Alcohol, 4.26% Polyethylene Glycol and 85.5% Nonreactive Ingredients
12/cs.
Videos
SDS
Documents

Documents

Promotions

Promotions