missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scopoletin, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 302900500

 View more versions of this product

Catalog No. AC302900500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Scopoletin
92-61-5
Beige
94% min. (HPLC)
C10H8O4
50 mg
scopoletin, gelseminic acid, 6-methylesculetin, 7-hydroxy-6-methoxy-2h-chromen-2-one, chrysatropic acid, scopoletine, murrayetin, scopoletol, 7-hydroxy-6-methoxycoumarin, 6-o-methylesculetin
COC1=C(C=C2C(=C1)C=CC(=O)O2)O
192.17
CHEBI:17488
95%
95%
200.0°C to 207.0°C
Authentic
Glass bottle
MFCD00006872
15, 8545
RODXRVNMMDRFIK-UHFFFAOYSA-N
7-hydroxy-6-methoxychromen-2-one
5280460
192.17
Crystalline Powder

Chemical Identifiers

92-61-5
192.17
RODXRVNMMDRFIK-UHFFFAOYSA-N
5280460
7-hydroxy-6-methoxychromen-2-one
C10H8O4
MFCD00006872
scopoletin, gelseminic acid, 6-methylesculetin, 7-hydroxy-6-methoxy-2h-chromen-2-one, chrysatropic acid, scopoletine, murrayetin, scopoletol, 7-hydroxy-6-methoxycoumarin, 6-o-methylesculetin
CHEBI:17488
COC1=C(C=C2C(=C1)C=CC(=O)O2)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-171-9

RTECSNumber : GN6930000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only